Merck
  • Carbonic anhydrase activators: activation of the beta-carbonic anhydrases from the pathogenic fungi Candida albicans and Cryptococcus neoformans with amines and amino acids.
Carbonic anhydrase activators: activation of the beta-carbonic anhydrases from the pathogenic fungi Candida albicans and Cryptococcus neoformans with amines and amino acids.
Bioorganic & medicinal chemistry (2010-01-12)
Alessio Innocenti, Rebecca A Hall, Andrea Scozzafava, Fritz A Mühlschlegel, Claudiu T Supuran
ABSTRACT
The proteins encoded by the Nce103 genes of Candida albicans and Cryptococcus neoformans are catalytically active beta-carbonic anhydrases (CAs, EC 4.2.1.1) playing various roles in the life cycle of these fungal pathogens, such as CO(2) sensing, regulation of capsule biosynthesis, filamentation, and adaptation of the organism to various pH and CO(2) conditions in various niches where the fungi grow. Here, we report the first activation study of these two enzymes, CaNce103 and Can2, respectively, with amines and amino acids. The C. albicans enzyme, CaNce103 was activated by amino acids such as L-/D-His, L-D-Trp, L-Tyr with K(A)s in the range of 19.5-46 microM. More effective activators were some amines such as histamine, dopamine, 2-aminoethyl-piperazine, and L-adrenaline (K(A)s of 13.2-18.5 microM). The best CaNce103 activators were L- and D-Dopa, with K(A)s of 0.96-2.5 microM. The C. neoformans enzyme, Can2, showed much lower propensity to be activated by all these amino acids and amines, which had activation constants in the range of 28.7-47.2 microM. The best Can2 activator was L-Trp. This study may help to better understand the catalytic/activation mechanisms of the beta-CAs and eventually to design CA activity modulators of such widespread enzymes in pathogenic fungi.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
SAFC
L-Tyrosine
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland