Merck
  • MymA Bioactivated Thioalkylbenzoxazole Prodrug Family Active against Mycobacterium tuberculosis.
MymA Bioactivated Thioalkylbenzoxazole Prodrug Family Active against Mycobacterium tuberculosis.
Journal of medicinal chemistry (2020-04-11)
Abraham L Moure, Gagandeep Narula, Flavia Sorrentino, Adama Bojang, Clement K M Tsui, Carine Sao Emani, Esther Porras-De Francisco, Beatriz Díaz, María José Rebollo-López, Pedro Alfonso Torres-Gómez, Eva María López-Román, Isabel Camino, Patricia Casado Castro, Laura Guijarro López, Fátima Ortega, Lluis Ballell, David Barros-Aguirre, Modesto Remuiñán Blanco, Yossef Av-Gay
ABSTRACT
Screening of a GSK-proprietary library against intracellular Mycobacterium tuberculosis identified 1, a thioalkylbenzoxazole hit. Biological profiling and mutant analysis revealed that this compound is a prodrug that is bioactivated by the mycobacterial enzyme MymA. A hit-expansion program including design, synthesis, and profiling of a defined set of analogues with optimized drug-like properties led to the identification of an emerging lead compound, displaying potency against intracellular bacteria in the low micromolar range, high in vitro solubility and permeability, and excellent microsomal stability.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture