Merck
  • Molecular properties of psychopharmacological drugs determining non-competitive inhibition of 5-HT3A receptors.
Molecular properties of psychopharmacological drugs determining non-competitive inhibition of 5-HT3A receptors.
European journal of medicinal chemistry (2009-01-16)
Johannes Kornhuber, Lothar Terfloth, Stefan Bleich, Jens Wiltfang, Rainer Rupprecht
ABSTRACT
We developed a structure-property-activity relationship (SPAR)-model for psychopharmacological drugs acting as non-competitive 5-HT(3A) receptor antagonists by using a decision-tree learner provided by the RapidMiner machine learning tool. A single molecular descriptor, namely the molecular dipole moment per molecular weight (mu/MW), predicts whether or not a substance non-competitively antagonizes 5-HT-induced Na(+) currents. A low mu/MW is compatible with drug-cumulation in apolar lipid rafts. This study confirms that size-intensive descriptors allow the development of compact SPAR models.
Sigma-Aldrich
Acridine, ≥96.5% (HPLC)
Sigma-Aldrich
10,11-Dihydro-5H-dibenz[b,f]azepine, 97%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder