Merck
Conditioned mate-guarding behavior in the female rat.
Physiology & behavior (2014-04-29)
Amanda Holley, Shy Shalev, Shannon Bellevue, James G Pfaus
ABSTRACT
Female and male rats are often described as having a promiscuous mating strategy, yet simple Pavlovian conditioning paradigms, in which a neutral odor or strain-related cues are paired with preferred sexual reward states during an animal's first sexual experiences, shift this strategy toward copulatory and mate preferences for partners bearing the familiar odor or strain cue. We examined whether female rats given exclusive rewarding copulation with one particular male would display mate-guarding behavior, a strong index of monogamous mating. Ovariectomized, hormone-primed female Long-Evans rats were given their first 10 paced sexual experiences at 4-day intervals with a particular unscented male of the same strain. A final test was conducted in an open field 4-days later in which the primed, partnered female was given access to the male partner and a fully-primed competitor female. In this situation, the partnered females mounted the competitor female repeatedly if she came near the vicinity of the male. This behavior prevented the male from copulating with the competitor, and was not displayed if partnered females could not pace the rate of copulatory behavior efficiently during the training trials, nor was it displayed by the competitor females. Fos expression was examined in both the partnered and competitor females after the final open field test. Partnered females had significantly higher expression within the supraoptic nucleus and nucleus accumbens shell compared to partnered females that did not develop this behavior or competitor females. These data show that females engaged in paced copulation with the same male display mate-guarding when exposed to that male and a competitor female. Increased activation of the SON and NAc may underlie this behavior.
Sigma-Aldrich
LSD1 Inhibitor IV, RN-1, 2HCl, LSD1 Inhibitor IV, RN-1, HCl, is a cell-permeable potent, irreversible inhibitor of lysine specific demethylase 1 (LSD1; IC₅₀ = 70 nM in a HRP-coupled assay using H3K4Me2 peptide substrate).
Sigma-Aldrich
Anti-Vasopressin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Oxytocin Antibody, serum, Chemicon®