Merck
  • Identification of the skeletal progenitor cells forming osteophytes in osteoarthritis.
Identification of the skeletal progenitor cells forming osteophytes in osteoarthritis.
Annals of the rheumatic diseases (2020-09-24)
Anke J Roelofs, Karolina Kania, Alexandra J Rafipay, Meike Sambale, Stephanie T Kuwahara, Fraser L Collins, Joanna Smeeton, Maxwell A Serowoky, Lynn Rowley, Hui Wang, René Gronewold, Chrysa Kapeni, Simón Méndez-Ferrer, Christopher B Little, John F Bateman, Thomas Pap, Francesca V Mariani, Joanna Sherwood, J Gage Crump, Cosimo De Bari
ABSTRACT
Osteophytes are highly prevalent in osteoarthritis (OA) and are associated with pain and functional disability. These pathological outgrowths of cartilage and bone typically form at the junction of articular cartilage, periosteum and synovium. The aim of this study was to identify the cells forming osteophytes in OA. Fluorescent genetic cell-labelling and tracing mouse models were induced with tamoxifen to switch on reporter expression, as appropriate, followed by surgery to induce destabilisation of the medial meniscus. Contributions of fluorescently labelled cells to osteophytes after 2 or 8 weeks, and their molecular identity, were analysed by histology, immunofluorescence staining and RNA in situ hybridisation. Pdgfrα-H2BGFP mice and Pdgfrα-CreER mice crossed with multicolour Confetti reporter mice were used for identification and clonal tracing of mesenchymal progenitors. Mice carrying Col2-CreER, Nes-CreER, LepR-Cre, Grem1-CreER, Gdf5-Cre, Sox9-CreER or Prg4-CreER were crossed with tdTomato reporter mice to lineage-trace chondrocytes and stem/progenitor cell subpopulations. Articular chondrocytes, or skeletal stem cells identified by Nes, LepR or Grem1 expression, did not give rise to osteophytes. Instead, osteophytes derived from Pdgfrα-expressing stem/progenitor cells in periosteum and synovium that are descendants from the Gdf5-expressing embryonic joint interzone. Further, we show that Sox9-expressing progenitors in periosteum supplied hybrid skeletal cells to the early osteophyte, while Prg4-expressing progenitors from synovial lining contributed to cartilage capping the osteophyte, but not to bone. Our findings reveal distinct periosteal and synovial skeletal progenitors that cooperate to form osteophytes in OA. These cell populations could be targeted in disease modification for treatment of OA.
Product Description
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
Anti-Fluorescein-POD, Fab fragments, from sheep
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Roche
Fluorescein RNA Labeling Mix, solution, suitable for Northern blotting, suitable for Southern blotting, suitable for hybridization