Merck
  • Synthesis of celecoxib analogues possessing a N-difluoromethyl-1,2-dihydropyrid-2-one 5-lipoxygenase pharmacophore: biological evaluation as dual inhibitors of cyclooxygenases and 5-lipoxygenase with anti-inflammatory activity.
Synthesis of celecoxib analogues possessing a N-difluoromethyl-1,2-dihydropyrid-2-one 5-lipoxygenase pharmacophore: biological evaluation as dual inhibitors of cyclooxygenases and 5-lipoxygenase with anti-inflammatory activity.
Journal of medicinal chemistry (2009-03-20)
Morshed A Chowdhury, Khaled R A Abdellatif, Ying Dong, Dipankar Das, Mavanur R Suresh, Edward E Knaus
ABSTRACT
A novel class of 1-(4-methanesulfonylphenyl and 4-aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole hybrid cyclooxygenase-2 (COX-2)/5-lipoxygenase (5-LOX) inhibitory anti-inflammatory agents was designed. Replacement of the tolyl ring present in celecoxib by the N-difluoromethyl-1,2-dihydropyrid-2-one moiety provided compounds showing dual selective COX-2/5-LOX inhibitory activities. 1-(4-Aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole exhibited good anti-inflammatory (AI) activity (ED(50) = 27.7 mg/kg po) that compares favorably with the reference drugs celecoxib (ED(50) = 10.8 mg/kg po) and ibuprofen (ED(50) = 67.4 mg/kg po). The N-difluoromethyl-1,2-dihydropyridin-2-one moiety provides a novel 5-LOX pharmacophore for the design of cyclic hydroxamic mimetics for exploitation in the development of COX-2/5-LOX inhibitory AI drugs.
Product Description
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications