Merck
  • Evolutionary Divergence of Enzymatic Mechanisms for Tubulin Detyrosination.
Evolutionary Divergence of Enzymatic Mechanisms for Tubulin Detyrosination.
Cell reports (2019-12-19)
Siem van der Laan, Maude F Lévêque, Guillaume Marcellin, Lubomir Vezenkov, Yoann Lannay, Geronimo Dubra, Guillaume Bompard, Sara Ovejero, Serge Urbach, Andrew Burgess, Muriel Amblard, Yvon Sterkers, Patrick Bastien, Krzysztof Rogowski
ABSTRACT
The two related members of the vasohibin family, VASH1 and VASH2, encode human tubulin detyrosinases. Here we demonstrate that, in contrast to VASH1, which requires binding of small vasohibin binding protein (SVBP), VASH2 has autonomous tubulin detyrosinating activity. Moreover, we demonstrate that SVBP acts as a bona fide activator of both enzymes. Phylogenetic analysis of the vasohibin family revealed that regulatory diversification of VASH-mediated tubulin detyrosination coincided with early vertebrate evolution. Thus, as a model organism for functional analysis, we used Trypanosoma brucei (Tb), an evolutionarily early-branched eukaryote that possesses a single VASH and encodes a terminal tyrosine on both α- and β-tubulin tails, both subject to removal. Remarkably, although detyrosination levels are high in the flagellum, TbVASH knockout parasites did not present any noticeable flagellar abnormalities. In contrast, we observed reduced proliferation associated with profound morphological and mitotic defects, underscoring the importance of tubulin detyrosination in cell division.
Sigma-Aldrich
Anti-SVBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Anti-Vasohibin-2, clone 5E3 Antibody, clone 5E3, 1 mg/mL, from mouse
Sigma-Aldrich
Anti-acetyl-alpha tubulin Antibody, clone 6-11B-1, clone 6-11B-1, from mouse
Sigma-Aldrich
Carboxypeptidase A from bovine pancreas, (Type II-PMSF treated), ≥50 units/mg protein, ready-to-use solution
Sigma-Aldrich
Anti-Vasohibin-1 Antibody, clone 4E12 (Azide Free), clone 4E12, 1 mg/mL, from mouse