Merck
  • Effects of doxorubicin associated with amniotic membrane stem cells in the treatment of canine inflammatory breast carcinoma (IPC-366) cells.
Effects of doxorubicin associated with amniotic membrane stem cells in the treatment of canine inflammatory breast carcinoma (IPC-366) cells.
BMC veterinary research (2020-09-26)
Jéssica Borghesi, Sara Caceres, Lara Carolina Mario, Angela Alonso-Diez, Ana Carolina Silveira Rabelo, Maria J Illera, Gema Silvan, Maria Angélica Miglino, Phelipe O Favaron, Ana Claudia O Carreira, Juan Carlos Illera
ABSTRACT
Tumours in mammary glands represent the most common neoplasia in bitches, as in humans. This high incidence results in part from the stimulation of sex hormones on these glands. Among mammary tumours, inflammatory carcinoma is the most aggressive, presenting a poor prognosis to surgical treatment and chemotherapy. One of the most widely used chemotherapy drugs for breast cancer treatment is doxorubicin (DOXO). Alternative therapies have been introduced in order to assist in these treatments; studies on treatments using stem cells have emerged, since they have anti-inflammatory and immunomodulatory properties. The aim of this study was to evaluate the effects of DOXO and canine amniotic membrane stem cells (AMCs) on the triple-negative canine inflammatory mammary carcinoma cell line IPC-366. Four experimental groups were analysed: a control group without treatment; Group I with DOXO, Group II with AMC and Group III with an association of DOXO and AMCs. We performed the MTT assay with DOXO in order to select the best concentration for the experiments. The growth curve was performed with all groups (I-III) in order to verify the potential of treatments to reduce the growth of IPC-366. For the cell cycle, all groups (I-III) were tested using propidium iodide. While in the flow cytometry, antibodies to progesterone receptor (PR), estrogen receptor (ER), PCNA, VEGF, IL-10 and TGF-β1 were used. For steroidogenic pathway hormones, an ELISA assay was performed. The results showed that cells treated with 10 µg/mL DOXO showed a 71.64% reduction in cellular growth after 72 h of treatment. Reductions in the expression of VEGF and PCNA-3 were observed by flow cytometry in all treatments when compared to the control. The intracellular levels of ERs were also significantly increased in Group III (4.67% vs. 27.1%). Regarding to the levels of steroid hormones, significant increases in the levels of estradiol (E2) and estrone sulphate (S04E1) were observed in Groups I and III. On the other hand, Group II did not show differences in steroid hormone levels in relation to the control. We conclude that the association of DOXO with AMCs (Group III) promoted a reduction in cell growth and in the expression of proteins related to proliferation and angiogenesis in IPC-366 triple-negative cells. This treatment promoted ER positive expression, suggesting that the accumulated oestrogen conducted these cells to a synergistic state, rendering these tumour cells responsive to ERs and susceptible to new hormonal cancer therapies.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Corning® CellBIND® Multiple Well Plate, size 6 wells, flat clear bottom, sterile, lid
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture