Merck
  • Critical role of histone H3 lysine 27 demethylase Kdm6b in the homeostasis and function of medullary thymic epithelial cells.
Critical role of histone H3 lysine 27 demethylase Kdm6b in the homeostasis and function of medullary thymic epithelial cells.
Cell death and differentiation (2020-04-30)
Zhi Liu, Haohao Zhang, Yiming Hu, Dandan Liu, Lingling Li, Cuifeng Li, Qi Wang, Junhaohui Huo, Hanshao Liu, Ningxia Xie, Xingxu Huang, Yongzhong Liu, Charlie Degui Chen, Yufang Shi, Xiaoren Zhang
ABSTRACT
Medullary thymic epithelial cells (mTECs) play a central role in the establishment of T cell central immunological tolerance by promiscuously expressing tissue-restricted antigens (TRAs) and presenting them to developing T cells, leading to deletion of T cells responding to self-antigens. However, molecular mechanisms especially epigenetic regulation of mTEC homeostasis and TRA expression remain elusive. Here we show that the H3K27 demethylase Kdm6b is essential to maintain the postnatal thymic medulla by promoting mTEC survival and regulating the expression of TRA genes. Moreover, mice lacking Kdm6b developed pathological autoimmune disorders. Mechanically, Kdm6b exerted its function by reducing repressive H3K27 trimethylation (H3K27me3) at the promoters of anti-apoptotic gene Bcl2 and a set of Aire-dependent TRA genes. Thus, our findings reveal a dual role of Kdm6b in the regulation of mTEC-mediated T cell central tolerance.
Sigma-Aldrich
SeqPlex DNA Amplification Kit, For use with high throughput sequencing technologies, Whole Genome Amplification kit designed to facilitate Next Gen Sequencing.
Sigma-Aldrich
Lectin from Ulex europaeus (gorse, furze), FITC conjugate, lyophilized powder
Sigma-Aldrich
2′-Deoxyguanosine monohydrate, powder, BioReagent, suitable for cell culture, 99-100%
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Millipore
Polycarbonate Membrane Filter, 0.8 μm Pore Size, Isopore, filter diam. 142 mm, hydrophilic