Merck
  • Targeting FTO Suppresses Cancer Stem Cell Maintenance and Immune Evasion.
Targeting FTO Suppresses Cancer Stem Cell Maintenance and Immune Evasion.
Cancer cell (2020-06-13)
Rui Su, Lei Dong, Yangchan Li, Min Gao, Li Han, Mark Wunderlich, Xiaolan Deng, Hongzhi Li, Yue Huang, Lei Gao, Chenying Li, Zhicong Zhao, Sean Robinson, Brandon Tan, Ying Qing, Xi Qin, Emily Prince, Jun Xie, Hanjun Qin, Wei Li, Chao Shen, Jie Sun, Prakash Kulkarni, Hengyou Weng, Huilin Huang, Zhenhua Chen, Bin Zhang, Xiwei Wu, Mark J Olsen, Markus Müschen, Guido Marcucci, Ravi Salgia, Ling Li, Amir T Fathi, Zejuan Li, James C Mulloy, Minjie Wei, David Horne, Jianjun Chen
ABSTRACT
Fat mass and obesity-associated protein (FTO), an RNA N6-methyladenosine (m6A) demethylase, plays oncogenic roles in various cancers, presenting an opportunity for the development of effective targeted therapeutics. Here, we report two potent small-molecule FTO inhibitors that exhibit strong anti-tumor effects in multiple types of cancers. We show that genetic depletion and pharmacological inhibition of FTO dramatically attenuate leukemia stem/initiating cell self-renewal and reprogram immune response by suppressing expression of immune checkpoint genes, especially LILRB4. FTO inhibition sensitizes leukemia cells to T cell cytotoxicity and overcomes hypomethylating agent-induced immune evasion. Our study demonstrates that FTO plays critical roles in cancer stem cell self-renewal and immune evasion and highlights the broad potential of targeting FTO for cancer therapy.
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Methoxy poly(ethylene glycol)-b-poly(D,L-lactide), 4k-2.2k
Sigma-Aldrich
Pyronin Y, for NA electrophoresis
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
D-Luciferin potassium salt, ≥98.0% (HPLC)
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 5,000