- Aberrant epigenetic regulation of bromodomain BRD4 in human colon cancer.
Aberrant epigenetic regulation of bromodomain BRD4 in human colon cancer.
Journal of molecular medicine (Berlin, Germany) (2011-11-29)
R M Rodriguez, C Huidobro, R G Urdinguio, C Mangas, B Soldevilla, G Domínguez, F Bonilla, A F Fernandez, M F Fraga
PMID22120039
ABSTRACT
The bromodomain protein BRD4 is involved in cell proliferation and cell cycle progression, primarily through its role in acetylated chromatin-dependent regulation of transcription at targeted loci. Here, we show that BRD4 is frequently downregulated by aberrant promoter hypermethylation in human colon cancer cell lines and primary tumors. Ectopic re-expression of BRD4 in these colon cancer cell lines markedly reduced in vivo tumor growth, suggesting a role of BRD4 in human colon cancer.
Sigma-Aldrich
BRD4 (342-460) His tag human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Sigma-Aldrich
BRD4, BD1, BD2 (49-460) His tag human, recombinant, expressed in E. coli, ≥75% (SDS-PAGE)
Sigma-Aldrich
BRD4 (342-460) GST tag human, recombinant, expressed in E. coli, ≥75% (SDS-PAGE)
Sigma-Aldrich
BRD4, BD1, BD2 (49-460) GST tag human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)