Merck
  • Photophysics of J-Aggregating Porphyrin-Lipid Photosensitizers in Liposomes: Impact of Lipid Saturation.
Photophysics of J-Aggregating Porphyrin-Lipid Photosensitizers in Liposomes: Impact of Lipid Saturation.
Langmuir : the ACS journal of surfaces and colloids (2020-04-28)
Danielle M Charron, Goonay Yousefalizadeh, Hilde H Buzzá, Maneesha A Rajora, Juan Chen, Kevin G Stamplecoskie, Gang Zheng
ABSTRACT
Porphyrin aggregates have attractive photophysical properties for phototherapy and optical imaging, including quenched photosensitization, efficient photothermal conversion, and unique absorption spectra. Although hydrophobic porphyrin photosensitizers have long been encapsulated into liposomes for drug delivery, little is known about the membrane properties of liposomes with large amphiphilic porphyrin compositions. In this paper, a porphyrin-lipid conjugate was incorporated into liposomes formed of saturated or unsaturated lipids to study the membrane composition-dependent formation of highly ordered porphyrin J-aggregates and disordered aggregates. Porphyrin-lipid readily phase-separates in saturated membranes, forming J-aggregates that are destabilized during the ripple phase below the main thermal transition. Porphyrin-lipid J-aggregates are photostable with a photothermal efficiency of 54 ± 6%, comparable to gold. Even at high porphyrin-lipid compositions, porphyrin J-aggregates coexist with a minority population of disordered aggregates, which are photodynamically active despite being fluorescently quenched. For photothermal applications, liposome formulations that encourage porphyrin-lipid phase separation should be explored for maximum J-aggregation.
Product Description
Avanti
18:0 Lyso PC, 1-stearoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
18:0 Lyso PC, Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:1 Lyso PC, Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand
Avanti
18:1 Lyso PC, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform