Merck
  • Phosphoprotein Detection with a Single Nanofluidic Diode Decorated with Zinc Chelates.
Phosphoprotein Detection with a Single Nanofluidic Diode Decorated with Zinc Chelates.
ChemPlusChem (2020-03-28)
Saima Nasir, Mubarak Ali, Ishtiaq Ahmed, Christof M Niemeyer, Wolfgang Ensinger
ABSTRACT
We report a nanofluidic device for the label-free detection of phosphoprotein (PPn) analytes. To achieve this goal, a metal ion chelator, namely 4-[bis(2-pyridylmethyl)aminomethyl]aniline (DPA-NH2 ) compound was synthesized. Single asymmetric nanofluidic channels were fabricated in polyethylene terephthalate (PET) membranes. The chelator (DPA-NH2 ) molecules are subsequently immobilized on the nanochannel surface, followed by the zinc ion complexation to afford DPA-Zn2+ chelates, which act as ligand moieties for the specific binding of phosphoproteins. The success of the chemical reaction and biomolecular recognition process that occur in a confined geometry can be monitored from the changes in electrical readout of the nanochannel. The nanofluidic sensor has the ability to sensitively and specifically detect lower concentrations (≥1 nM) of phosphoprotein (albumin and α-casein) in the surrounding environment as evidenced from the significant decrease in ion current flowing through the nanochannels. However, dephosphoproteins such as lysozyme and dephospho-α-casein even at higher concentration (>1 μM) could not induce any significant change in the transmembrane ion flux. This observation indicated the sensitivity and specificity of the proposed nanofluidic sensor towards PPn proteins, and has potential for use in differentiating between phosphoproteins and dephosphoproteins.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
α-Casein from bovine milk, ≥70% αs-casein basis (electrophoresis), lyophilized powder
Sigma-Aldrich
α-Casein Dephosphorylated from bovine milk, lyophilized powder
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein