Merck
Deacetylation Inhibition Reverses PABPN1-Dependent Muscle Wasting.
iScience (2019-02-11)
Cyriel S Olie, Muhammad Riaz, Rebecca Konietzny, Philip D Charles, Adan Pinto-Fernandez, Szymon M Kiełbasa, A Aartsma-Rus, Jelle J Goeman, Benedikt M Kessler, Vered Raz
ABSTRACT
Reduced poly(A)-binding protein nuclear 1 (PABPN1) levels cause aging-associated muscle wasting. PABPN1 is a multifunctional regulator of mRNA processing. To elucidate the molecular mechanisms causing PABPN1-mediated muscle wasting, we compared the transcriptome with the proteome in mouse muscles expressing short hairpin RNA to PABPN1 (shPab). We found greater variations in the proteome than in mRNA expression profiles. Protein accumulation in the shPab proteome was concomitant with reduced proteasomal activity. Notably, protein acetylation appeared to be decreased in shPab versus control proteomes (63%). Acetylome profiling in shPab muscles revealed prominent peptide deacetylation associated with elevated sirtuin-1 (SIRT1) deacetylase. We show that SIRT1 mRNA levels are controlled by PABPN1 via alternative polyadenylation site utilization. Most importantly, SIRT1 deacetylase inhibition by sirtinol increased PABPN1 levels and reversed muscle wasting. We suggest that perturbation of a multifactorial regulatory loop involving PABPN1 and SIRT1 plays an imperative role in aging-associated muscle wasting. VIDEO ABSTRACT.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-20S proteasome subunit β5 Antibody, serum, Upstate®