Merck
  • Increased ceramide content and NFκB signaling may contribute to the attenuation of anabolic signaling after resistance exercise in aged males.
Increased ceramide content and NFκB signaling may contribute to the attenuation of anabolic signaling after resistance exercise in aged males.
Journal of applied physiology (Bethesda, Md. : 1985) (2012-10-09)
Donato A Rivas, Evan P Morris, Prashanth H Haran, Evan P Pasha, Mauricio da Silva Morais, Gregory G Dolnikowski, Edward M Phillips, Roger A Fielding
ABSTRACT
One of the most fundamental adaptive physiological events is the response of skeletal muscle to high-intensity resistance exercise, resulting in increased protein synthesis and ultimately larger muscle mass. However, muscle growth in response to contraction is attenuated in older humans. Impaired contractile-induced muscle growth may contribute to sarcopenia: the age-associated loss of muscle mass and function that is manifested by loss of strength, contractile capacity, and endurance. We hypothesized that the storage of ceramide would be increased in older individuals and this would be associated with increases in NFκB signaling and a decreased anabolic response to exercise. To test this hypothesis we measured ceramides at rest and anabolic and NFκB signaling after an acute bout of high-intensity resistance exercise in young and older males. Using lipidomics analysis we show there was a 156% increase in the accumulation of C16:0-ceramide (P < 0.05) and a 30% increase in C20:0-ceramide (P < 0.05) in skeletal muscle with aging, although there was no observable difference in total ceramide. C16:0-ceramide content was negatively correlated (P = 0.008) with lower leg lean mass. Aging was associated with a ~60% increase in the phosphorylation of the proinflammatory transcription factor NFκB in the total and nuclear cell fractions (P < 0.05). Furthermore, there was an attenuated activation of anabolic signaling molecules such as Akt (P < 0.05), FOXO1 (P < 0.05), and S6K1 (P < 0.05) after an acute bout of high-intensity resistance exercise in older males. We conclude that ceramide may have a significant role in the attenuation of contractile-induced skeletal muscle adaptations and atrophy that is observed with aging.
Product Description
Avanti
C18 Ceramide (d18:1/18:0), Avanti Research - A Croda Brand
Avanti
C24 Ceramide-1-Phosphate (d18:1/24:0), Avanti Research - A Croda Brand 860527P, powder
Avanti
C14 Ceramide (d18:1/14:0), Avanti Research - A Croda Brand 860514P, powder
Avanti
C24 Ceramide (d18:1/24:0), Avanti Research - A Croda Brand
Avanti
C20 Ceramide (d18:1/20:0), Avanti Research - A Croda Brand 860520P, powder
Avanti
C16 Ceramide (d18:1/16:0), Avanti Research - A Croda Brand
Avanti
C24:1 Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand
Avanti
C18:1 Ceramide (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860519P, powder
Avanti
Sphingosine (d18:1), Avanti Research - A Croda Brand
Avanti
C22 Ceramide (d18:1/22:0), Avanti Research - A Croda Brand 860501P, powder
Avanti
sphingosine-d7, Avanti Research - A Croda Brand 860657P, powder