Merck
  • Construction of a liposome dialyzer for the preparation of high-value, small-volume liposome formulations.
Construction of a liposome dialyzer for the preparation of high-value, small-volume liposome formulations.
Nature protocols (2015-05-29)
Katarzyna Adamala, Aaron E Engelhart, Neha P Kamat, Lin Jin, Jack W Szostak
ABSTRACT
The liposome dialyzer is a small-volume equilibrium dialysis device, built from commercially available materials, that is designed for the rapid exchange of small volumes of an extraliposomal reagent pool against a liposome preparation. The dialyzer is prepared by modification of commercially available dialysis cartridges (Slide-A-Lyzer cassettes), and it consists of a reactor with two 300-μl chambers and a 1.56-cm(2) dialysis surface area. The dialyzer is prepared in three stages: (i) disassembling the dialysis cartridges to obtain the required parts, (ii) assembling the dialyzer and (iii) sealing the dialyzer with epoxy. Preparation of the dialyzer takes ∼1.5 h, not including overnight epoxy curing. Each round of dialysis takes 1-24 h, depending on the analyte and membrane used. We previously used the dialyzer for small-volume non-enzymatic RNA synthesis reactions inside fatty acid vesicles. In this protocol, we demonstrate other applications, including removal of unencapsulated calcein from vesicles, remote loading and vesicle microscopy.
Product Description
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer