Merck
  • Genomic Decoding of Neuronal Depolarization by Stimulus-Specific NPAS4 Heterodimers.
Genomic Decoding of Neuronal Depolarization by Stimulus-Specific NPAS4 Heterodimers.
Cell (2019-10-05)
G Stefano Brigidi, Michael G B Hayes, Nathaniel P Delos Santos, Andrea L Hartzell, Lorane Texari, Pei-Ann Lin, Anna Bartlett, Joseph R Ecker, Christopher Benner, Sven Heinz, Brenda L Bloodgood
ABSTRACT
Cells regulate gene expression in response to salient external stimuli. In neurons, depolarization leads to the expression of inducible transcription factors (ITFs) that direct subsequent gene regulation. Depolarization encodes both a neuron's action potential (AP) output and synaptic inputs, via excitatory postsynaptic potentials (EPSPs). However, it is unclear if distinct types of electrical activity can be transformed by an ITF into distinct modes of genomic regulation. Here, we show that APs and EPSPs in mouse hippocampal neurons trigger two spatially segregated and molecularly distinct induction mechanisms that lead to the expression of the ITF NPAS4. These two pathways culminate in the formation of stimulus-specific NPAS4 heterodimers that exhibit distinct DNA binding patterns. Thus, NPAS4 differentially communicates increases in a neuron's spiking output and synaptic inputs to the nucleus, enabling gene regulation to be tailored to the type of depolarizing activity along the somato-dendritic axis of a neuron.
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Oligo(dT)23, Anchored, 70 μM in H2O
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
Millipore
NP-40 Alternative
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 20%, for molecular biology
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
SpeedBeads magnetic carboxylate modified particles, Cytiva 65152105050250, 0.70-1.10 μm avg. part. size, suspension (5% Solids)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture