Merck
  • microRNA-301b-3p downregulation underlies a novel inhibitory role of long non-coding RNA MBNL1-AS1 in non-small cell lung cancer.
microRNA-301b-3p downregulation underlies a novel inhibitory role of long non-coding RNA MBNL1-AS1 in non-small cell lung cancer.
Stem cell research & therapy (2019-05-23)
Peng Li, Wenqun Xing, Jinliang Xu, Dongfeng Yuan, Guanghui Liang, Baoxing Liu, Haibo Ma
ABSTRACT
Non-small cell lung cancer (NSCLC) is the second most prevalent cause of cancer-related fatality. Long non-coding RNAs (lncRNAs) have been observed to exercise functions in NSCLC. Here, the current study aimed to explore the potential mechanism of lncRNA MBNL1-AS1 in NSCLC. Microarray analysis was performed to screen the differentially expressed lncRNA associated with NSCLC and its potential mechanism. The lncRNA MBNL1-AS1 expression was quantified in 56 paired NSCLC and adjacent normal tissue samples. In an attempt to outline the function of lncRNA MBNL1-AS1 in NSCLC and to identify the interaction among lncRNA MBNL1-AS1, microRNA-301b-3p (miR-301b-3p) and TGFBR2, ectopic expression, depletion, and reporter assay experiments were conducted to detect CSC proliferation, migration, invasion, drug resistance, and sphere formation in NSCLC. Initially, the intersection among lncRNA MBNL1-AS1, miR-301b-3p, and TGFBR2 was observed in NSCLC. While a poor expression of lncRNA MBNL1-AS1 and TGFBR2, along with a high expression of miR-301b-3p was observed in NSCLC tissues. A demonstration of lncRNA MBNL1-AS1 restoration significantly decreased CSC proliferation, migration, invasion, drug resistance, and sphere formation in NSCLC. LncRNA MBNL1-AS1 functioned as a sponge of miR-301b-3p, which inverted the inhibitory role of lncRNA MBNL1-AS1 in CSC proliferation, migration, invasion, drug resistance, and sphere formation in NSCLC. LncRNA MBNL1-AS1 positively regulated TGFBR2 which was a target gene of miR-301b-3p. At last, upregulated lncRNA MBNL1-AS1 or depleted miR-301b-3p suppressed the xenograft tumor formation in vivo. Collectively, the present study suggests an inhibitory role of lncRNA MBNL1-AS1 in CSC drug resistance of NSCLC by upregulating miR-301b-3p-targeted TGFBR2.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Gefitinib, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-POU5F1 (Oct4) antibody produced in mouse, ~1.0 mg/mL, clone PF-29, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-SMAD2 (pThr220) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Smad3 (pSer204) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-ABCG2 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
S(−)-Verapamil hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-TGFBR2 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-SMAD3 antibody produced in rabbit