Merck
  • Magnolin protects against contrast-induced nephropathy in rats via antioxidation and antiapoptosis.
Magnolin protects against contrast-induced nephropathy in rats via antioxidation and antiapoptosis.
Oxidative medicine and cellular longevity (2014-11-18)
Feng Wang, Guangyuan Zhang, Yang Zhou, Dingkun Gui, Junhui Li, Tao Xing, Niansong Wang
ABSTRACT
Magnolin is the major active ingredient of the herb Magnolia fargesii which has anti-inflammatory and antioxidative effects. Oxidative stress and apoptosis are involved in the pathogenesis of contrast-induced nephropathy (CIN). We hypothesize that Magnolin could protect against CIN through antioxidative and antiapoptotic properties. To test whether Magnolin could attenuate CIN, oxidative stress and apoptosis, in vivo and in vitro, we utilized a rat model of ioversol-induced CIN and a cell model of oxidative stress in which HK2 cells were treated with H2O2. Rats were assigned to 4 groups (n = 6 per group): control group, ioversol group (ioversol-induced CIN), vehicle group (CIN rats pretreated with vehicle), and Magnolin group (CIN rats pretreated with 1 mg/kg Magnolin). The results showed that magnolin ameliorated the renal tubular necrosis, apoptosis, and the deterioration of renal function (P < 0.05). Furthermore, Magnolin reduced the renal oxidative stress, suppressed caspase-3 activity, and increased Bcl-2 expression in vivo and in vitro. Magnolin might protect CIN in rats through antioxidation and antiapoptosis.
Sigma-Aldrich
Magnolin, ≥95% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder