Merck
  • Determination of electrostatic potentials at biological interfaces using electron-electron double resonance.
Determination of electrostatic potentials at biological interfaces using electron-electron double resonance.
Biophysical journal (1992-06-01)
Y K Shin, W L Hubbell
ABSTRACT
A new general method for the determination of electrostatic potentials at biological surfaces is presented. The approach is based on measurement of the collision frequency of a charged nitroxide in solution with a nitroxide fixed to the surface at the point of interest. The collision frequency is determined with 14N:15N double label electron-electron double resonance (ELDOR). As a test, the method is shown to give values for phospholipid bilayer surface potentials consistent with the Gouy-Chapman theory, a simple model shown by many independent tests to accurately describe charged, planar surfaces. In addition, the method is applied to determine the electrostatic potential near the surface of DNA. The results indicate that the potential is significantly smaller than that predicted from Poisson-Boltzmann analysis, but is in qualitative agreement with that predicted by Manning's theory of counter ion condensation. The method is readily extended to measurement of surface potentials of proteins.
Product Description
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
4-palmitamido-TEMPO, N-tempoyl palmitamide, powder