Merck
  • Incorporating allylated lignin-derivatives in thiol-ene gel-polymer electrolytes.
Incorporating allylated lignin-derivatives in thiol-ene gel-polymer electrolytes.
International journal of biological macromolecules (2018-03-06)
Elyse A Baroncini, Joseph F Stanzione
ABSTRACT
Growing environmental and economic concerns as well as the uncertainty that accompanies finite petrochemical resources contributes to the increase in research and development of bio-based, renewable polymers. Concurrently, industrial and consumer demand for smaller, safer, and more flexible technologies motivates a global research effort to improve electrolytic polymer separators in lithium-ion batteries. To incorporate the aromatic structural advantages of lignin, a highly abundant and renewable resource, into gel-polymer electrolytes, lignin-derived molecules, vanillyl alcohol and gastrodigenin are functionalized and UV-polymerized with multi-functional thiol monomers. The resulting thin, flexible, polymer films possess glass transition temperatures ranging from -42.1°C to 0.3°C and storage moduli at 25°C ranging from 1.90MPa to 10.08MPa. The crosslinked polymer films swollen with electrolyte solution impart conductivities in the range of 7.04×10-7 to 102.73×10-7Scm-1. Thiol molecular weight has the most impact on the thermo-mechanical properties of the resulting films while polymer crosslink density has the largest effect on conductivity. The conducting abilities of the bio-based gel-polymer electrolytes in this study prove the viability of lignin-derived feedstock for use in lithium-ion battery applications and reveal structurally and thermally desirable traits for future work.
Sigma-Aldrich
Allyl bromide, ReagentPlus®, 99%, contains ≤1000 ppm propylene oxide as stabilizer
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%
Sigma-Aldrich
3-Methoxybenzyl alcohol, 98%
Sigma-Aldrich
Benzyltriethylammonium chloride, 99%