Merck
  • Evolutionary novelty in gravity sensing through horizontal gene transfer and high-order protein assembly.
Evolutionary novelty in gravity sensing through horizontal gene transfer and high-order protein assembly.
PLoS biology (2018-04-25)
Tu Anh Nguyen, Jamie Greig, Asif Khan, Cara Goh, Gregory Jedd
ABSTRACT
Horizontal gene transfer (HGT) can promote evolutionary adaptation by transforming a species' relationship to the environment. In most well-understood cases of HGT, acquired and donor functions appear to remain closely related. Thus, the degree to which HGT can lead to evolutionary novelties remains unclear. Mucorales fungi sense gravity through the sedimentation of vacuolar protein crystals. Here, we identify the octahedral crystal matrix protein (OCTIN). Phylogenetic analysis strongly supports acquisition of octin by HGT from bacteria. A bacterial OCTIN forms high-order periplasmic oligomers, and inter-molecular disulphide bonds are formed by both fungal and bacterial OCTINs, suggesting that they share elements of a conserved assembly mechanism. However, estimated sedimentation velocities preclude a gravity-sensing function for the bacterial structures. Together, our data suggest that HGT from bacteria into the Mucorales allowed a dramatic increase in assembly scale and emergence of the gravity-sensing function. We conclude that HGT can lead to evolutionary novelties that emerge depending on the physiological and cellular context of protein assembly.
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
L-(+)-Ergothioneine
Sigma-Aldrich
Monoclonal Anti-mCherry antibody produced in mouse, clone GT235, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody