
Valproic Acid Related Compound B

Synonym(s):
2-Isopropylpentanoic acid, 2-Isopropyl-2-propylacetic acid
Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2