Skip to Content
Merck
Trimethyl orthovalerate 97%

Trimethyl orthovalerate

Trimethyl orthovalerate 97%
Synonym(s):
1,1,1-Trimethoxypentane
Linear Formula:
CH3CH2CH2CH2C(OCH3)3
CAS Number:
Molecular Weight:
162.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing