
Trazodone Related Compound C

Synonym(s):
2-{3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl}-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
Empirical Formula (Hill Notation):
C19H22ClN5O · HCl
CAS Number:
Molecular Weight:
408.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2