
trans-β-Nitrostyrene

Synonym(s):
trans-1-Nitro-2-phenylethylene
Linear Formula:
C6H5CH=CHNO2
CAS Number:
Molecular Weight:
149.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing