
Tranexamic acid Related Compound C

Synonym(s):
(RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic acid
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2