
Topotecan hydrochloride

Synonym(s):
(S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione monohydrochloride, NSC 609669
Empirical Formula (Hill Notation):
C23H23N3O5 · HCl
CAS Number:
Molecular Weight:
457.91
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing