
Timolol Related Compound D

Synonym(s):
4-Morpholino-1,2,5-thiadiazol-3-ol
Empirical Formula (Hill Notation):
C6H9N3O2S
CAS Number:
Molecular Weight:
187.22
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2