Skip to Content
Merck
SUPELCOGEL™ C610H, 6% Crosslinked HPLC Column 9 μm particle size, L × I.D. 30 cm × 7.8 mm

SUPELCOGEL C610 (9 µm) HPLC Columns

SUPELCOGEL™ C610H, 6% Crosslinked HPLC Column 9 μm particle size, L × I.D. 30 cm × 7.8 mm
Synonym(s):
SUPELCOGEL 6% Cross Linked HPLC Columns

Products

Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing