
Sodium tetrafluoroborate

Synonym(s):
Sodium fluoroborate, Sodium tetrafluoroborate
Linear Formula:
NaBF4
CAS Number:
Molecular Weight:
109.79
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-3 of 3