
Silica gel on TLC Al foils

Synonym(s):
TLC cards Silica gel
Products
Product Category
Brand
Manufacturer
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing