
Selegiline hydrochloride

Synonym(s):
R-(−)-Deprenyl hydrochloride, (R)-(−)-N,α-Dimethyl-N-(2-propynyl)phenethylamine hydrochloride, R(−)-N-α-Dimethyl-N-2-propynyl-benzeneethanamine hydrochloride, Selegiline hydrochloride
Empirical Formula (Hill Notation):
C13H17N · HCl
CAS Number:
Molecular Weight:
223.74
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing