
Rhoifolin

Synonym(s):
Apigenin 7-O-neohesperidoside, Rhoifolin, Rhoifoloside
Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing