
Pyridoxine

Synonym(s):
Pyridoxol, Vitamin B6
Empirical Formula (Hill Notation):
C8H11NO3
CAS Number:
Molecular Weight:
169.18
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reference Material Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing