
Pregnenolone sulfate sodium salt

Synonym(s):
3β-Hydroxy-5-pregnen-20-one 3-sulfate, 3-Hydroxypregn-5-en-20-one sulfate, 5-Pregnen-3β-ol-20-one sulfate
Empirical Formula (Hill Notation):
C21H31NaO5S
Molecular Weight:
418.52
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing