
Pravastatin Related Compound A

Synonym(s):
(βR,δR,1S,2R,3S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-β,δ,3-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoic acid monosodium salt
Empirical Formula (Hill Notation):
C23H35NaO7
CAS Number:
Molecular Weight:
446.51
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2