
Potassium palmitate-13C16

Synonym(s):
Hexadecanoic acid-13C16 potassium salt, Palmitic acid-13C16 potassium salt, Potassium hexadecanoate-13C16
Linear Formula:
13CH3(13CH2)1413CO2K
CAS Number:
Molecular Weight:
310.40
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing