
Poly(styrene)-block-poly(acrylic acid), azide terminated

Synonym(s):
PS-b-PAA, Polystyrene-polyacrylic acid polymersome forming polymer
Linear Formula:
CH3CH(C6H5)[CH2CH(C6H5)]x [CH2CH(CO2H)]y N3
Products
Brand
Melting Point (°C)
Physical Form
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing