
Poly(N-isopropylacrylamide)

Synonym(s):
NIPAM polymer, PNIPAM, Polyacrylamide, polyNIPAM
Linear Formula:
CH3(C6H11NO)nCH3
CAS Number:
Products
Brand
Melting Point (°C)
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing