
Polylactide-block-poly(ethylene glycol)-block-polylactide

Synonym(s):
PLA-PEG-PLA
Linear Formula:
HO(CHCH3COO)x(CH2CH2O)y(COCHCH3O)zH
Products
Brand
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing