
Poly(ethylene glycol) methyl ether methacrylate

Synonym(s):
Methoxy PEG methacrylate, Methoxy poly(ethylene glycol) monomethacrylate, Poly(ethylene glycol) monomethyl ether monomethacrylate
Linear Formula:
H2C=CCH3CO2(CH2CH2O)nCH3
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Showing 1-4 of 4
Compare
Product Number
Description
Pricing