Skip to Content
Merck
Poly(ethylene glycol) bisazide average Mn 20,000

Poly(ethylene glycol) bisazide

Poly(ethylene glycol) bisazide average Mn 20,000
Synonym(s):
Polyoxyethylene bis(azide), O,O′-Bis(2-azidoethyl)polyethylene glycol, PEG-diazide
Linear Formula:
N3CH2CH2(OCH2CH2)nN3
CAS Number:

Products

Brand
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Showing 1-2 of 2