Piperidine, polymer-bound
Synonym(s):
(Piperidinomethyl)polystyrene, 1-Benzylpiperidine polymer-bound, Copolymer of styrene and divinylbenzene, piperidinomethylated, Polystyrene crosslinked with divinylbenzene, piperidinomethylated
Products
Product Category
Brand
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing