Skip to Content
Merck
Phosphoric acid-d3 solution 85 wt. % in D2O, 98 atom % D

Phosphoric acid-d3 solution

Phosphoric acid-d3 solution 85 wt. % in D2O, 98 atom % D
Synonym(s):
Trideuterophosphoric acid
Linear Formula:
D3PO4
CAS Number:
Molecular Weight:
101.01

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing