
Phenylephrine Related Compound D

Synonym(s):
(R-3-{2-[Benzyl(methyl)amino]-1-hydroxyethyl}phenol
Empirical Formula (Hill Notation):
C16H19NO2
CAS Number:
Molecular Weight:
257.33
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2