
Perphenazine for system suitability

Synonym(s):
Perphenazine, 4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol
Empirical Formula (Hill Notation):
C21H26ClN3OS
CAS Number:
Molecular Weight:
403.97
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing