
Perfluoroheptanoic acid

Synonym(s):
Tridecafluoroheptanoic acid, Perfluoroheptanoic acid
Linear Formula:
CF3(CF2)5CO2H
CAS Number:
Molecular Weight:
364.06
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3