Skip to Content
Merck
[Pd(acac)2] Umicore, 99%

[Pd(acac)2]

[Pd(acac)2] Umicore, 99%
Synonym(s):
Palladium(II) acetylacetonate, Palladium(II) 2,4-pentanedionate, Pd(acac)2
Linear Formula:
Pd(C5H7O2)2
CAS Number:
Molecular Weight:
304.64
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing