![O-(2-Mercaptoethyl)-O′-methylpolyethylene glycol 10,000](/deepweb/assets/sigmaaldrich/product/structures/157/329/e09b5e4f-8871-41c8-989f-4fd2a4c42e17/640/e09b5e4f-8871-41c8-989f-4fd2a4c42e17.png)
O-(2-Mercaptoethyl)-O′-methylpolyethylene glycol
![O-(2-Mercaptoethyl)-O′-methylpolyethylene glycol 10,000](/deepweb/assets/sigmaaldrich/product/structures/157/329/e09b5e4f-8871-41c8-989f-4fd2a4c42e17/640/e09b5e4f-8871-41c8-989f-4fd2a4c42e17.png)
Synonym(s):
MeO-PEG-SH, Mercaptopolyethylene glycol monomethyl ether, PEG-thiol, Polyethylene glycol 2-mercaptoethyl methyl ether
CAS Number:
Products
Brand
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing